Learn More
Meso Scale Discovery U-PLEX Human VEGF-a ASYQP
SDP
Catalog No. 5000078296
Encompass
The U-PLEX Human VEGF-A Assay (K151UVK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of vascular endothelial growth factor A in human biological samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable VEGF-A quantification in research applications
Catalog No. 50-000-78296 Supplier Meso Scale Discovery Supplier No. K151UVK21
The U-PLEX Human VEGF-A Assay (K151UVK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of vascular endothelial growth factor A in human biological samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable VEGF-A quantification in research applications