Learn More
Meso Scale Discovery U-PLEX Human VEGF-D ASY
SDP
Catalog No. 5000082016
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay for measuring VEGF-D in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The assay quantifies VEGF-D as low as 0 3 pg/mL with 25 L of sample It includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals The kit is specific with no cross-reactivity allowing rapid quantification of VEGF-D in 3 5 hours
Catalog No. 50-000-82016 Supplier Meso Scale Discovery Supplier No. K151C8K1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay for measuring VEGF-D in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The assay quantifies VEGF-D as low as 0 3 pg/mL with 25 L of sample It includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals The kit is specific with no cross-reactivity allowing rapid quantification of VEGF-D in 3 5 hours