Learn More
Meso Scale Discovery U-PLEX Human VEGF-D ASYQP
SDP
Catalog No. 5000082029
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay that measures VEGF-D levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify VEGF-D concentrations as low as 0 05 pg/mL with as little as 25 L of sample It includes a 96-well plate pre-coated with capture antibodies detection antibodies read buffer wash buffer calibrator diluent and a vial of VEGF-D calibrator The kit enables accurate reproducible quantitation of VEGF-D levels in about 3 5 hours
Catalog No. 50-000-82029 Supplier Meso Scale Discovery Supplier No. K151C8K21
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay that measures VEGF-D levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify VEGF-D concentrations as low as 0 05 pg/mL with as little as 25 L of sample It includes a 96-well plate pre-coated with capture antibodies detection antibodies read buffer wash buffer calibrator diluent and a vial of VEGF-D calibrator The kit enables accurate reproducible quantitation of VEGF-D levels in about 3 5 hours