Learn More
Meso Scale Discovery U-PLEX Human vWF ASY
SDP
Catalog No. 5000078334
Encompass
The Meso Scale Discovery U-PLEX Human VWF Assay Kit measures von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluent wash buffer read buffer and plate seals Results are available in 3 5 hours using an MSD instrument offering a rapid high-throughput method for VWF measurement in research
Catalog No. 50-000-78334 Supplier Meso Scale Discovery Supplier No. K151C9K2
The Meso Scale Discovery U-PLEX Human VWF Assay Kit measures von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluent wash buffer read buffer and plate seals Results are available in 3 5 hours using an MSD instrument offering a rapid high-throughput method for VWF measurement in research