Learn More
Meso Scale Discovery U-PLEX Human vWF ASYQP
SDP
Catalog No. 5000078500
Encompass
The Meso Scale Discovery U-PLEX Human VWF Assay is an electrochemiluminescent immunoassay for quantifying von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology with a detection limit of 0 03 ng/mL and quantification up to 100 ng/mL Requiring 50 L of sample the assay takes about 3 5 hours It aids in diagnosing von Willebrand disease and other VWF-related conditions The kit includes a 96-well plate detection antibody calibrators diluent and necessary reagents
Catalog No. 50-000-78500 Supplier Meso Scale Discovery Supplier No. K151C9K21
The Meso Scale Discovery U-PLEX Human VWF Assay is an electrochemiluminescent immunoassay for quantifying von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology with a detection limit of 0 03 ng/mL and quantification up to 100 ng/mL Requiring 50 L of sample the assay takes about 3 5 hours It aids in diagnosing von Willebrand disease and other VWF-related conditions The kit includes a 96-well plate detection antibody calibrators diluent and necessary reagents