Learn More
Meso Scale Discovery U-PLEX Human vWF ASYQP
SDP
Catalog No. 5000082063
Encompass
The Meso Scale Discovery U-PLEX Human VWF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of VWF with a wide dynamic range The assay requires a small sample volume and has a total assay time of approximately 3 5 hours The lower limit of detection is 1 3 ng/mL and the upper limit of quantitation is 10 000 ng/mL This assay provides a simple rapid high-throughput method for measuring VWF levels
Catalog No. 50-000-82063 Supplier Meso Scale Discovery Supplier No. K151C9K22
The Meso Scale Discovery U-PLEX Human VWF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of VWF with a wide dynamic range The assay requires a small sample volume and has a total assay time of approximately 3 5 hours The lower limit of detection is 1 3 ng/mL and the upper limit of quantitation is 10 000 ng/mL This assay provides a simple rapid high-throughput method for measuring VWF levels