Learn More
Meso Scale Discovery U-PLEX Human YKL-40 ASY
SDP
Catalog No. 5000078220
Encompass
The Meso Scale Discovery U-PLEX Human YKL-40 Assay Kit is an electrochemiluminescent immunoassay for quantifying YKL-40 in human serum plasma and cell culture supernatants It uses the SECTOR Imager 6000 for high sensitivity and a wide dynamic range The kit includes a 96-well plate detection antibody calibrators and diluent It detects as low as 1 15 pg/mL and up to 100 000 pg/mL requires 50 L sample volume and has 10% variation The assay completes in about 3 5 hours
Catalog No. 50-000-78220 Supplier Meso Scale Discovery Supplier No. K151VLK2
The Meso Scale Discovery U-PLEX Human YKL-40 Assay Kit is an electrochemiluminescent immunoassay for quantifying YKL-40 in human serum plasma and cell culture supernatants It uses the SECTOR Imager 6000 for high sensitivity and a wide dynamic range The kit includes a 96-well plate detection antibody calibrators and diluent It detects as low as 1 15 pg/mL and up to 100 000 pg/mL requires 50 L sample volume and has 10% variation The assay completes in about 3 5 hours