Learn More
Meso Scale Discovery U-PLEX Human YKL-40 ASY
SDP
Catalog No. 5000081835
Encompass
The Meso Scale Discovery U-PLEX Human YKL-40 Assay Kit is a multiplex immunoassay that quantitatively measures YKL-40 protein levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer sufficient for one 96-well plate Recommended applications include biomarker research and monitoring therapeutic response
Catalog No. 50-000-81835 Supplier Meso Scale Discovery Supplier No. K151VLK1
The Meso Scale Discovery U-PLEX Human YKL-40 Assay Kit is a multiplex immunoassay that quantitatively measures YKL-40 protein levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer sufficient for one 96-well plate Recommended applications include biomarker research and monitoring therapeutic response