Learn More
Meso Scale Discovery U-PLEX Human YKL-40 ASYQP
SDP
Catalog No. 5000081424
Encompass
The U-PLEX Human YKL-40 Assay (K151VLK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of YKL-40 biomarker in human samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes calibrators controls and detection reagents optimized for serum plasma and cell culture samples providing reliable results for research applications in inflammation and disease biomarker studies
Catalog No. 50-000-81424 Supplier Meso Scale Discovery Supplier No. K151VLK22
The U-PLEX Human YKL-40 Assay (K151VLK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of YKL-40 biomarker in human samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes calibrators controls and detection reagents optimized for serum plasma and cell culture samples providing reliable results for research applications in inflammation and disease biomarker studies