Learn More
Meso Scale Discovery U-PLEX Metabolic Combo 1 ms
SDP
Catalog No. 5000080016
Encompass
The U-PLEX Metabolic Combo 1 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin leptin resistin and PAI-1 in mouse serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to run the assay Results are obtained in as little as 3 5 hours from a small sample volume The kit has high lot-to-lot consistency and is suitable for mouse metabolic research
Catalog No. 50-000-80016 Supplier Meso Scale Discovery Supplier No. K15297K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Combo 1 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin leptin resistin and PAI-1 in mouse serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to run the assay Results are obtained in as little as 3 5 hours from a small sample volume The kit has high lot-to-lot consistency and is suitable for mouse metabolic research