Learn More
Meso Scale Discovery U-PLEX Metabolic Combo 1 ms
SDP
Catalog No. 5000082100
Encompass
The U-PLEX Metabolic Combo 1 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin leptin resistin and PAI-1 in mouse serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology provides high sensitivity and a broad dynamic range requires low sample volume and has a short 2 5 hour total assay time The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents sufficient for 1 x 96-well plate Recommended for research use only
Catalog No. 50-000-82100 Supplier Meso Scale Discovery Supplier No. K15297K1
The U-PLEX Metabolic Combo 1 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin leptin resistin and PAI-1 in mouse serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology provides high sensitivity and a broad dynamic range requires low sample volume and has a short 2 5 hour total assay time The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents sufficient for 1 x 96-well plate Recommended for research use only