Learn More
Meso Scale Discovery U-PLEX Metabolic Combo 1 rt
SDP
Catalog No. 5000082161
Encompass
The U-PLEX Metabolic Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of insulin leptin and resistin in rat serum plasma and tissue culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay allows sensitive detection of the analytes using an MSD instrument The kit has a broad dynamic range and provides accurate and reproducible results with as little as 25 L sample
Catalog No. 50-000-82161 Supplier Meso Scale Discovery Supplier No. K15308K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of insulin leptin and resistin in rat serum plasma and tissue culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay allows sensitive detection of the analytes using an MSD instrument The kit has a broad dynamic range and provides accurate and reproducible results with as little as 25 L sample