Learn More
Meso Scale Discovery U-PLEX Metabolic Combo 2 hu
SDP
Catalog No. 5000079798
Encompass
The U-PLEX Metabolic Combo 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of insulin leptin PAI-1 active resistin and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent assay takes approximately 3 5 hours to complete and has a detection range of 56 6-50 000 pg/mL for insulin 23 4-25 000 pg/mL for leptin 1 37-1500 pg/mL for PAI-1 active 15 6-25 000 pg/mL for resistin and 1 64-2000 pg/mL for TNF-alpha
Catalog No. 50-000-79798 Supplier Meso Scale Discovery Supplier No. K15388K1
The U-PLEX Metabolic Combo 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of insulin leptin PAI-1 active resistin and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent assay takes approximately 3 5 hours to complete and has a detection range of 56 6-50 000 pg/mL for insulin 23 4-25 000 pg/mL for leptin 1 37-1500 pg/mL for PAI-1 active 15 6-25 000 pg/mL for resistin and 1 64-2000 pg/mL for TNF-alpha