Learn More
Meso Scale Discovery U-PLEX Metabolic Combo 2 hu
SDP
Catalog No. 5000082084
Encompass
The U-PLEX Metabolic Combo 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of insulin leptin PAI-1 active resistin and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay takes approximately 4 hours to complete and has a detection range of 0 05-2500 pg/mL across the analytes The kit includes reagents for one 96-well plate and can test 40 samples in duplicate
Catalog No. 50-000-82084 Supplier Meso Scale Discovery Supplier No. K15388K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Combo 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of insulin leptin PAI-1 active resistin and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay takes approximately 4 hours to complete and has a detection range of 0 05-2500 pg/mL across the analytes The kit includes reagents for one 96-well plate and can test 40 samples in duplicate