Learn More
Meso Scale Discovery U-PLEX Mouse 6CKine/CCL21 ASY
SDP
Catalog No. 5000082210
Encompass
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay kit for measuring 6 mouse cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IFN- CCL21) from various samples It uses MSD s electrochemiluminescent detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay takes about 5 hours and includes all necessary reagents and plates for 96 tests suitable for mouse samples only aiding in immunology and inflammation research
Catalog No. 50-000-82210 Supplier Meso Scale Discovery Supplier No. K152C0K2
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay kit for measuring 6 mouse cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IFN- CCL21) from various samples It uses MSD s electrochemiluminescent detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay takes about 5 hours and includes all necessary reagents and plates for 96 tests suitable for mouse samples only aiding in immunology and inflammation research