Learn More
Meso Scale Discovery U-PLEX Mouse BCA-1/BLC ASY
SDP
Catalog No. 5000081329
Encompass
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay Kit is a multiplex immunoassay for quantifying BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents It provides insights into immune function and inflammation enabling efficient and accurate biomarker analysis
Catalog No. 50-000-81329 Supplier Meso Scale Discovery Supplier No. K152F0K1
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay Kit is a multiplex immunoassay for quantifying BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents It provides insights into immune function and inflammation enabling efficient and accurate biomarker analysis