Learn More
Meso Scale Discovery U-PLEX Mouse BCA-1/BLC ASYQP
SDP
Catalog No. 5000079256
Encompass
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay is a multiplex immunoassay kit for measuring BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection from a small sample with results in 3 5 hours It requires an MSD instrument like MESO QuickPlex SQ 120 for reading With minimal sample needs and quick results this kit is efficient for mouse BCA-1 and BLC quantification in research
Catalog No. 50-000-79256 Supplier Meso Scale Discovery Supplier No. K152F0K22
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay is a multiplex immunoassay kit for measuring BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection from a small sample with results in 3 5 hours It requires an MSD instrument like MESO QuickPlex SQ 120 for reading With minimal sample needs and quick results this kit is efficient for mouse BCA-1 and BLC quantification in research