Learn More
Meso Scale Discovery U-PLEX Mouse BCA-1/BLC ASYQP
SDP
Catalog No. 5000081453
Encompass
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay is a multiplex immunoassay kit for measuring BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation This assay efficiently analyzes BCA-1 and BLC biomarkers crucial for immune function and inflammation
Catalog No. 50-000-81453 Supplier Meso Scale Discovery Supplier No. K152F0K21
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay is a multiplex immunoassay kit for measuring BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation This assay efficiently analyzes BCA-1 and BLC biomarkers crucial for immune function and inflammation