Learn More
Meso Scale Discovery U-PLEX Mouse C-Peptide ASY
SDP
Catalog No. 5000082075
Encompass
The Meso Scale Discovery C-Peptide Kit is an electrochemiluminescent immunoassay for measuring C-peptide in human serum plasma and other samples It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity wide range and quick results Detection limits are 26 pg/mL to 20 000 pg/mL with a sample volume of 25 L The assay takes 3 5 hours The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators controls buffers and seals For research use only
Catalog No. 50-000-82075 Supplier Meso Scale Discovery Supplier No. K1526JK1
The Meso Scale Discovery C-Peptide Kit is an electrochemiluminescent immunoassay for measuring C-peptide in human serum plasma and other samples It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity wide range and quick results Detection limits are 26 pg/mL to 20 000 pg/mL with a sample volume of 25 L The assay takes 3 5 hours The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators controls buffers and seals For research use only