Learn More
Meso Scale Discovery U-PLEX Mouse C-Peptide ASY
SDP
Catalog No. 5000082176
Encompass
The Meso Scale Discovery U-PLEX Mouse C-Peptide Assay is a highly sensitive immunoassay that quantitatively measures C-peptide levels in mouse serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides accurate and reproducible results with an ultra-low detection limit of 0 006 ng/mL The assay requires only 25 L of sample and can be run on MSD s SECTOR Imager 6000 for high throughput screening This product enables the sensitive and accurate measurement of C-peptide levels which serves as a marker of insulin secretion and -cell function
Catalog No. 50-000-82176 Supplier Meso Scale Discovery Supplier No. K1526JK2
The Meso Scale Discovery U-PLEX Mouse C-Peptide Assay is a highly sensitive immunoassay that quantitatively measures C-peptide levels in mouse serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides accurate and reproducible results with an ultra-low detection limit of 0 006 ng/mL The assay requires only 25 L of sample and can be run on MSD s SECTOR Imager 6000 for high throughput screening This product enables the sensitive and accurate measurement of C-peptide levels which serves as a marker of insulin secretion and -cell function