Learn More
Meso Scale Discovery U-PLEX Mouse C-Peptide ASYQP
SDP
Catalog No. 5000078479
Encompass
The Meso Scale Discovery U-PLEX Mouse C-Peptide Assay is a multiplex immunoassay kit that measures C-peptide levels in mouse serum plasma and other biological samples It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay can be run on Meso Scale Discovery s SECTOR Imager instruments It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The kit provides reagents for testing 89 samples in duplicate It allows sensitive reproducible quantification of C-peptide levels from mouse samples
Catalog No. 50-000-78479 Supplier Meso Scale Discovery Supplier No. K1526JK22
The Meso Scale Discovery U-PLEX Mouse C-Peptide Assay is a multiplex immunoassay kit that measures C-peptide levels in mouse serum plasma and other biological samples It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay can be run on Meso Scale Discovery s SECTOR Imager instruments It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The kit provides reagents for testing 89 samples in duplicate It allows sensitive reproducible quantification of C-peptide levels from mouse samples