Learn More
Meso Scale Discovery U-PLEX Mouse CD40/TNFRSF5 ASY
SDP
Supplier:  Meso Scale Discovery K152D0K1
Encompass
The Meso Scale Discovery U-PLEX Mouse CD40 Assay is a multiplex immunoassay kit for quantifying mouse CD40 in various samples It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 The kit includes a plate with capture antibodies detection antibodies and necessary calibrators and controls for standard curve generation and assay validation enabling rapid and accurate mouse CD40 measurement
Catalog No. 50-000-81356