Learn More
Meso Scale Discovery U-PLEX Mouse CD40/TNFRSF5 ASY
SDP
Supplier:  Meso Scale Discovery K152D0K2
Encompass
The Meso Scale Discovery U-PLEX Mouse CD40 Assay Kit is a multiplex immunoassay for measuring CD40 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the QuickPlex SQ 120 The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrators for standard curve generation and assay validation enabling simultaneous detection of CD40 and other biomarkers
Catalog No. 50-000-81450