Learn More
Meso Scale Discovery U-PLEX Mouse Eotaxin ASY
SDP
Catalog No. 5000080436
Encompass
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that measures mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators ranging from 3 2 pg/mL to 10 000 pg/mL
Catalog No. 50-000-80436 Supplier Meso Scale Discovery Supplier No. K152UDK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that measures mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators ranging from 3 2 pg/mL to 10 000 pg/mL