Learn More
Meso Scale Discovery U-PLEX Mouse Eotaxin ASYQP
SDP
Catalog No. 5000078354
Encompass
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is designed to quantitatively measure mouse Eotaxin (CCL11) in serum plasma cell culture supernatants cell lysates tissue homogenates and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse Eotaxin The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse Eotaxin
Catalog No. 50-000-78354 Supplier Meso Scale Discovery Supplier No. K152UDK21
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is designed to quantitatively measure mouse Eotaxin (CCL11) in serum plasma cell culture supernatants cell lysates tissue homogenates and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse Eotaxin The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse Eotaxin