Learn More
Meso Scale Discovery U-PLEX Mouse Eotaxin ASYQP
SDP
Catalog No. 5000080439
Encompass
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that measures mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and buffer for sample dilution It can accurately quantify mouse Eotaxin concentrations with minimal hands-on time using an MSD instrument
Catalog No. 50-000-80439 Supplier Meso Scale Discovery Supplier No. K152UDK22
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that measures mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and buffer for sample dilution It can accurately quantify mouse Eotaxin concentrations with minimal hands-on time using an MSD instrument