Learn More
Meso Scale Discovery U-PLEX Mouse EPO ASY 25 PL
SDP
Catalog No. 5000082180
Encompass
The Meso Scale Discovery U-PLEX Mouse EPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse EPO with a lower limit of detection of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The assay can be run on MSD s SECTOR Imager 6000 for high throughput screening This quantitative immunoassay allows the accurate measurement of mouse EPO levels for the study of erythropoiesis regulation
Catalog No. 50-000-82180 Supplier Meso Scale Discovery Supplier No. K152VXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse EPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse EPO with a lower limit of detection of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The assay can be run on MSD s SECTOR Imager 6000 for high throughput screening This quantitative immunoassay allows the accurate measurement of mouse EPO levels for the study of erythropoiesis regulation