Learn More
Meso Scale Discovery U-PLEX Mouse EPO ASY384-well
SDP
Catalog No. 5000080182
Encompass
The Meso Scale Discovery U-PLEX Mouse EPO 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 14 pg/mL and an upper limit of quantification of 1000 pg/mL It requires 2 5 L sample size and can perform 38 samples in duplicate The total assay time is 3 5 hours This assay allows high-throughput sensitive and reproducible quantification of EPO from mouse samples
Catalog No. 50-000-80182 Supplier Meso Scale Discovery Supplier No. K252VXK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse EPO 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 14 pg/mL and an upper limit of quantification of 1000 pg/mL It requires 2 5 L sample size and can perform 38 samples in duplicate The total assay time is 3 5 hours This assay allows high-throughput sensitive and reproducible quantification of EPO from mouse samples