Learn More
Meso Scale Discovery U-PLEX Mouse EPO ASYQP
SDP
Catalog No. 5000078672
Encompass
MSD s EPO (erythropoietin) assays are electrochemiluminescence-based immunoassays designed for quantitative measurement of erythropoietin levels in biological samples These research-use-only kits utilize MULTI-SPOT and MULTI-ARRAY plate formats offering high sensitivity detection with minimal sample volumes The assays support drug development biomarker research and clinical studies requiring precise EPO quantification in serum plasma and other matrices
Catalog No. 50-000-78672 Supplier Meso Scale Discovery Supplier No. K152VXK22
MSD s EPO (erythropoietin) assays are electrochemiluminescence-based immunoassays designed for quantitative measurement of erythropoietin levels in biological samples These research-use-only kits utilize MULTI-SPOT and MULTI-ARRAY plate formats offering high sensitivity detection with minimal sample volumes The assays support drug development biomarker research and clinical studies requiring precise EPO quantification in serum plasma and other matrices