Learn More
Meso Scale Discovery U-PLEX Mouse FGF-21 ASY
SDP
Catalog No. 5000081405
Encompass
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is a multiplex immunoassay for measuring FGF-21 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL allowing simultaneous measurement of FGF-21 and other biomarkers
Catalog No. 50-000-81405 Supplier Meso Scale Discovery Supplier No. K1525WK1
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is a multiplex immunoassay for measuring FGF-21 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL allowing simultaneous measurement of FGF-21 and other biomarkers