Learn More
Meso Scale Discovery U-PLEX Mouse FGF-21 ASYQP
SDP
Catalog No. 5000078450
Encompass
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is a multiplex immunoassay for measuring mouse fibroblast growth factor 21 (FGF-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It aids research in metabolism diabetes obesity liver disease and FGF-21 biology
Catalog No. 50-000-78450 Supplier Meso Scale Discovery Supplier No. K1525WK21
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is a multiplex immunoassay for measuring mouse fibroblast growth factor 21 (FGF-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It aids research in metabolism diabetes obesity liver disease and FGF-21 biology