Learn More
Meso Scale Discovery U-PLEX Mouse FGF-21 ASYQP
SDP
Catalog No. 5000081350
Encompass
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of FGF-21 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies specific for mouse FGF-21 detection antibodies conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 5 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81350 Supplier Meso Scale Discovery Supplier No. K1525WK22
The Meso Scale Discovery U-PLEX Mouse FGF-21 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of FGF-21 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies specific for mouse FGF-21 detection antibodies conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 5 pg/mL and an upper limit of quantification of 10 000 pg/mL