Learn More
Meso Scale Discovery U-PLEX Mouse GLP-1 total ASY
SDP
Catalog No. 5000082171
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 5 pg/mL The assay measures active GLP-1 as well as the primary metabolite GLP-1 (9-36) amide It is for research use only
Catalog No. 50-000-82171 Supplier Meso Scale Discovery Supplier No. K1525UK2
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 5 pg/mL The assay measures active GLP-1 as well as the primary metabolite GLP-1 (9-36) amide It is for research use only