Learn More
Meso Scale Discovery U-PLEX Mouse Glucagon ASYQP
SDP
Catalog No. 5000079381
Encompass
The Meso Scale Discovery U-PLEX Mouse Glucagon Assay Kit is a multiplex immunoassay that allows simultaneous measurement of glucagon in mouse serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent The assay has a typical sensitivity of 0 26 pg/mL and an upper limit of quantification of 10 000 pg/mL for glucagon It requires only 25 L of sample per well and the total assay time is 3 5 hours
Catalog No. 50-000-79381 Supplier Meso Scale Discovery Supplier No. K1525YK21
The Meso Scale Discovery U-PLEX Mouse Glucagon Assay Kit is a multiplex immunoassay that allows simultaneous measurement of glucagon in mouse serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent The assay has a typical sensitivity of 0 26 pg/mL and an upper limit of quantification of 10 000 pg/mL for glucagon It requires only 25 L of sample per well and the total assay time is 3 5 hours