Learn More
Meso Scale Discovery U-PLEX Mouse GM-CSF ASY
SDP
Catalog No. 5000078714
Encompass
The Meso Scale Discovery U-PLEX Mouse GM-CSF Assay Kit is a multiplex immunoassay for measuring mouse GM-CSF in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture and detection antibodies calibrators and controls It provides high specificity no matrix interference and results in under 3 5 hours with a detection limit of 0 04 pg/mL for accurate quantitation
Catalog No. 50-000-78714 Supplier Meso Scale Discovery Supplier No. K152UMK2
The Meso Scale Discovery U-PLEX Mouse GM-CSF Assay Kit is a multiplex immunoassay for measuring mouse GM-CSF in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture and detection antibodies calibrators and controls It provides high specificity no matrix interference and results in under 3 5 hours with a detection limit of 0 04 pg/mL for accurate quantitation