Learn More
Meso Scale Discovery U-PLEX Mouse GM-CSF ASY
SDP
Catalog No. 5000082155
Encompass
The Meso Scale Discovery U-PLEX Mouse GM-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse granulocyte-macrophage colony-stimulating factor (GM-CSF) across samples This assay utilizes MSD s proprietary QuickPlex SQ 120 instrument and U-PLEX technology allowing detection of proteins peptides and antibodies in diverse sample types With minimal sample volume requirements and rapid results in under 3 hours this kit provides sensitive reproducible quantitation of mouse GM-CSF with minimal hands-on time The U-PLEX Mouse GM-CSF Assay provides a streamlined workflow for mouse GM-CSF biomarker analysis
Catalog No. 50-000-82155 Supplier Meso Scale Discovery Supplier No. K152UMK1
The Meso Scale Discovery U-PLEX Mouse GM-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse granulocyte-macrophage colony-stimulating factor (GM-CSF) across samples This assay utilizes MSD s proprietary QuickPlex SQ 120 instrument and U-PLEX technology allowing detection of proteins peptides and antibodies in diverse sample types With minimal sample volume requirements and rapid results in under 3 hours this kit provides sensitive reproducible quantitation of mouse GM-CSF with minimal hands-on time The U-PLEX Mouse GM-CSF Assay provides a streamlined workflow for mouse GM-CSF biomarker analysis