Learn More
Meso Scale Discovery U-PLEX Mouse GM-CSF ASYQP
SDP
Catalog No. 5000082156
Encompass
MSD s U-PLEX Mouse GM-CSF assay is an electrochemiluminescent immunoassay for quantifying granulocyte-macrophage colony-stimulating factor in mouse samples This multiplex-compatible assay uses SULFO-TAG labels and carbon electrodes for sensitive specific detection The U-PLEX platform allows researchers to measure multiple biomarkers simultaneously in small sample volumes making it ideal for cytokine profiling studies The assay provides reliable quantification of GM-CSF levels in serum plasma and cell culture supernatants from mouse models
Catalog No. 50-000-82156 Supplier Meso Scale Discovery Supplier No. K152UMK21
MSD s U-PLEX Mouse GM-CSF assay is an electrochemiluminescent immunoassay for quantifying granulocyte-macrophage colony-stimulating factor in mouse samples This multiplex-compatible assay uses SULFO-TAG labels and carbon electrodes for sensitive specific detection The U-PLEX platform allows researchers to measure multiple biomarkers simultaneously in small sample volumes making it ideal for cytokine profiling studies The assay provides reliable quantification of GM-CSF levels in serum plasma and cell culture supernatants from mouse models