Learn More
Meso Scale Discovery U-PLEX Mouse IFN- ASY
SDP
Catalog No. 5000080420
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is designed to quantitatively measure mouse interferon alpha (IFN- ) in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse IFN- The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse IFN-
Catalog No. 50-000-80420 Supplier Meso Scale Discovery Supplier No. K152W1K1
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is designed to quantitatively measure mouse interferon alpha (IFN- ) in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse IFN- The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse IFN-