Learn More
Meso Scale Discovery U-PLEX Mouse IFN- ASYQP
SDP
Catalog No. 5000078688
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is a multiplex immunoassay that measures mouse interferon alpha in serum plasma cell culture supernatant and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for mouse IFN alpha detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for the quantitative measurement of mouse IFN alpha levels with accuracy and precision
Catalog No. 50-000-78688 Supplier Meso Scale Discovery Supplier No. K152W1K22
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is a multiplex immunoassay that measures mouse interferon alpha in serum plasma cell culture supernatant and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for mouse IFN alpha detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for the quantitative measurement of mouse IFN alpha levels with accuracy and precision