Learn More
Meso Scale Discovery U-PLEX Mouse IFN ASYQP
SDP
Catalog No. 5000079354
Encompass
The MSD U-PLEX Mouse IFN assay is an electrochemiluminescence-based immunoassay for quantifying interferon-gamma levels in mouse samples This multiplex platform enables researchers to measure cytokine concentrations with high sensitivity and precision using small sample volumes The U-PLEX technology allows for customizable panel creation and provides reliable reproducible results for immunology and inflammation research applications in mouse models
Catalog No. 50-000-79354 Supplier Meso Scale Discovery Supplier No. K152TTK22
The MSD U-PLEX Mouse IFN assay is an electrochemiluminescence-based immunoassay for quantifying interferon-gamma levels in mouse samples This multiplex platform enables researchers to measure cytokine concentrations with high sensitivity and precision using small sample volumes The U-PLEX technology allows for customizable panel creation and provides reliable reproducible results for immunology and inflammation research applications in mouse models