Learn More
Meso Scale Discovery U-PLEX Mouse IFN-B ASY
SDP
Catalog No. 5000078226
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN beta Assay is a multiplex immunoassay kit for detecting and quantifying mouse interferon beta in various samples This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results from 25 L of sample It includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluents buffers and seals The assay s sensitivity is 0 06 pg/mL with an upper quantification limit of 2500 pg/mL for IFN beta
Catalog No. 50-000-78226 Supplier Meso Scale Discovery Supplier No. K152G0K1
The Meso Scale Discovery U-PLEX Mouse IFN beta Assay is a multiplex immunoassay kit for detecting and quantifying mouse interferon beta in various samples This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results from 25 L of sample It includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluents buffers and seals The assay s sensitivity is 0 06 pg/mL with an upper quantification limit of 2500 pg/mL for IFN beta