Learn More
Meso Scale Discovery U-PLEX Mouse IFN-B ASYQP
SDP
Catalog No. 5000082225
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN beta Assay is a multiplex immunoassay kit for detecting and quantifying mouse interferon beta in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls buffers plate seals and a manual This assay supports high-throughput screening of IFN beta levels in immunology and neurobiology research
Catalog No. 50-000-82225 Supplier Meso Scale Discovery Supplier No. K152G0K21
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse IFN beta Assay is a multiplex immunoassay kit for detecting and quantifying mouse interferon beta in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls buffers plate seals and a manual This assay supports high-throughput screening of IFN beta levels in immunology and neurobiology research