Learn More
Meso Scale Discovery U-PLEX Mouse IFN-gamma Assay, 5 plates
SDP
Catalog No. NC1202783
Encompass
The U-PLEX Mouse IFN-gamma Assay provides the components for the quantitative determination of both natural and recombinant IFN-gamma in serum, plasma, and cell culture supernatants.
Catalog No. NC1202783 Supplier Meso Scale Discovery Supplier No. K152TTK2
The U-PLEX Mouse IFN-gamma Assay provides the components for the quantitative determination of both natural and recombinant IFN-gamma in serum, plasma, and cell culture supernatants.