Learn More
Meso Scale Discovery U-PLEX Mouse IL-10 ASY
SDP
Catalog No. 5000080438
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-10 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 10 (IL-10) in various samples It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay quantifies mouse IL-10 for immunology inflammation and cytokine research
Catalog No. 50-000-80438 Supplier Meso Scale Discovery Supplier No. K152TZK2
The Meso Scale Discovery U-PLEX Mouse IL-10 Assay Kit is a multiplex immunoassay for measuring mouse interleukin 10 (IL-10) in various samples It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay quantifies mouse IL-10 for immunology inflammation and cytokine research