Learn More
Meso Scale Discovery U-PLEX Mouse IL-10 ASY384-well
SDP
Catalog No. 5000081256
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-10 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 10 (IL-10) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision Designed for the MSD QUICKPLEX SQ 120 and MESO SECTOR S 600 instruments it includes a 384-well plate with capture antibodies detection antibodies and necessary calibrators and controls to quantify IL-10 levels enabling efficient and accurate measurement of this anti-inflammatory cytokine
Catalog No. 50-000-81256 Supplier Meso Scale Discovery Supplier No. K252TZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-10 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 10 (IL-10) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision Designed for the MSD QUICKPLEX SQ 120 and MESO SECTOR S 600 instruments it includes a 384-well plate with capture antibodies detection antibodies and necessary calibrators and controls to quantify IL-10 levels enabling efficient and accurate measurement of this anti-inflammatory cytokine