Learn More
Meso Scale Discovery U-PLEX Mouse IL-10 ASYQP
SDP
Catalog No. 5000082152
Encompass
The MSD U-PLEX Mouse IL-10 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-10 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-10 a key anti-inflammatory cytokine using MSD s proprietary MULTI-SPOT and MULTI-ARRAY plate technology for enhanced sensitivity and reproducibility in research applications
Catalog No. 50-000-82152 Supplier Meso Scale Discovery Supplier No. K152TZK22
Add to Cart
Add to Cart
The MSD U-PLEX Mouse IL-10 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-10 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-10 a key anti-inflammatory cytokine using MSD s proprietary MULTI-SPOT and MULTI-ARRAY plate technology for enhanced sensitivity and reproducibility in research applications