Learn More
Meso Scale Discovery U-PLEX Mouse IL-12p70 ASY
SDP
Catalog No. 5000082153
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 12 (IL-12) in serum plasma cell culture supernatants and other biological samples This assay kit utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity detection The kit includes a mouse IL-12p70-specific antibody coated plate and a ruthenium labeled mouse IL-12p70 antibody The assay has a detection range of 1 5-1000 pg/mL and exhibits no cross-reactivity with other cytokines This product can aid in the research of inflammatory and infectious diseases cancer and immune system disorders involving IL-12 signaling
Catalog No. 50-000-82153 Supplier Meso Scale Discovery Supplier No. K152UAK2
The Meso Scale Discovery U-PLEX Mouse IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 12 (IL-12) in serum plasma cell culture supernatants and other biological samples This assay kit utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity detection The kit includes a mouse IL-12p70-specific antibody coated plate and a ruthenium labeled mouse IL-12p70 antibody The assay has a detection range of 1 5-1000 pg/mL and exhibits no cross-reactivity with other cytokines This product can aid in the research of inflammatory and infectious diseases cancer and immune system disorders involving IL-12 signaling