Learn More
Meso Scale Discovery U-PLEX Mouse IL-12p70 ASYQP
SDP
Catalog No. 5000078708
Encompass
The U-PLEX Mouse IL-12p70 Assay (K152UAK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-12p70 levels in mouse biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for cytokine detection in research applications The kit provides reliable reproducible results for studying immune responses and inflammatory processes in mouse models making it valuable for immunology and biomedical research laboratories
Catalog No. 50-000-78708 Supplier Meso Scale Discovery Supplier No. K152UAK21
The U-PLEX Mouse IL-12p70 Assay (K152UAK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-12p70 levels in mouse biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for cytokine detection in research applications The kit provides reliable reproducible results for studying immune responses and inflammatory processes in mouse models making it valuable for immunology and biomedical research laboratories