Learn More
Meso Scale Discovery U-PLEX Mouse IL-13 ASY 25 PL
SDP
Catalog No. 5000079094
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-13 Assay Kit is a multiplex immunoassay that allows simultaneous measurement of mouse interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators controls 10X diluent read buffer wash buffer and adhesive plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse IL-13
Catalog No. 50-000-79094 Supplier Meso Scale Discovery Supplier No. K152UBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-13 Assay Kit is a multiplex immunoassay that allows simultaneous measurement of mouse interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators controls 10X diluent read buffer wash buffer and adhesive plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for mouse IL-13