Learn More
Meso Scale Discovery U-PLEX Mouse IL-13 ASY
SDP
Catalog No. 5000078437
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-13 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 13 (IL-13) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling This IL-13 assay delivers rapid results in just 3 5 hours
Catalog No. 50-000-78437 Supplier Meso Scale Discovery Supplier No. K152UBK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse IL-13 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 13 (IL-13) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling This IL-13 assay delivers rapid results in just 3 5 hours